Print…
Ready-to-Load™ kits, DNA-based screening for smallpox
Fornecedor: EDVOTEK
124
EDVO124EA 0 EUR
EDVO124
Ready-to-Load™ kits, DNA-based screening for smallpox
Materiais educationais Biology Educational Materials Genetics Learning Activities
This experiment presents a bioterrorism scenario, with students examining a simulated DNA fingerprinting test for the detection of smallpox.
  • For six lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Fornecimento: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 ml calibrated drop pipette, 100 ml graduated cyclinder and microtipped transfer pipettes.