Print…
TASKI Sani Calc Pur-Eco W3b Washroom Descaler
Fornecedor: Diversey
7515243
DIVY7515243EA 54.4 EUR
DIVY7515243
TASKI Sani Calc Pur-Eco W3b Washroom Descaler
Soluções de limpeza Detergentes e Limpadores de Superfícies
Powerful limescale remover for acid resistant hard surfaces.
  • Fast acting acidic, based on citric acid formula
  • Excellent cleaning performance
  • Strong descaling properties
  • No scrubbing necessary at high concentration
Certificações: EU Ecolabel (EU Flower).
Novo produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable