Print…
Detergent, Disinfectant, Oxivir® Plus
Fornecedor: Diversey
100827423 100827422
DIVY100827423EA 251 EUR
DIVY100827423 DIVY100827422
Detergent, Disinfectant, Oxivir® Plus
Soluções de limpeza Desinfectantes
Cleaner disinfectant that eliminates a broad spectrum of microorganisms (bacteria, viruses, mold fungi, yeasts). It utilises the environmentally friendly Accelerated Hydrogen Peroxide (AHP®) technology: When using the product it decomposes into water and oxygen.
  • Safe on all acid resistant surfaces including upholstery and carpets
  • Ideal for prevention of outbreaks and as part of a body spillage programme
  • Woolsafe approved for soft furnishings
Certificações: EN 14476 Norovirus in 2 minutes. EN 1276 E. Coli in 5 minutes. EN 1650 in 15Mais informação do produto