Print…
Anti-vibration pad, Richter
Fornecedor: DIVERSIFIED BIOTECH
VIBE-1000
DIVEVIBE-1000EA 110 EUR
DIVEVIBE-1000
Anti-vibration pad, Richter
Protectores de superfícies Protectores de Bancadas
Richter anti-vibration pad dampens vibrations generated from lab equipment. It creates a stable surface that will keep equipment in place.
  • Small, lightweight design makes transportation between benches and laboratories easy
  • Pads can be stacked for heavier units or laid side-by-side for use with larger equipment
Ideal for use with vortexes, shakers, rockers and microscopes.