Print…
Strips of 8 PCR tubes with attached flat cap
Fornecedor: DELTALAB
4096.2N
DELT4096.2NEA 434 EUR
DELT4096.2N
Strips of 8 PCR tubes with attached flat cap
Tubos Tubos para PCR
Made of PP.
  • RNAse, DNAse and pyrogen free
  • Excellent chemical resistance
  • Excellent temperature resistance
Every cap embodies a shield in order to prevent contamination when being opened. Positive sealing will fully protect the contents from evaporation during the whole thermal cycle. These strips can be cut to any length while each tube has its own attached cap.