Print…
Cryovials
Fornecedor: DELTALAB
409105.1 409106.1 409107 409108 409109
DELT409105.1EA 73.2 EUR
DELT409105.1 DELT409106.1 DELT409107 DELT409108 DELT409109
Cryovials
Vials
Tubes and caps made of autoclavable PP. Designed for the storage of biological material at temperatures as low as –196 °C (only in the gas state if stored in liquid nitrogen).
  • Printed graduations and white marking area for sample identification
  • Colour code inserts can be fitted into the tube caps for identification
  • Withstand centrifugation up to 17000×g
  • Certified RNAse, DNAse and pyrogen free
Fornecimento: Cryovials are supplied capped.
Atenção: Note: Skirted tubes are not recommended for centrifugation.