Print…
Sample container, with screw cap, EUROTUBO®
Fornecedor: DELTALAB
202845 202846 202847
DELT202845EA 155 EUR
DELT202845 DELT202846 DELT202847
Sample container, with screw cap, EUROTUBO®
Recipientes de amostras Recipientes, uso geral
PP, translucent.
  • With double sealed screw cap
  • Good chemical resistance
  • Good temperature resistance