Print…
Supelco® TraceCERT® Organic Reference Standard, Triazophos
Fornecedor: Merck
CRM74120-50MG
CRM74120-50MGEA 210 EUR
CRM74120-50MG
Supelco® TraceCERT® Organic Reference Standard, Triazophos
Padrões Organic Standards
Certificações: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Atenção: P260 - P264 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310