Print…
Digital vortex mixer, multiposition, Corning® LSE™
Fornecedor: Corning
6710
CORN6710EA 1280.88 EUR
CORN6710
Digital vortex mixer, multiposition, Corning® LSE™
Agitadores Agitadores Vortex
The Corning® LSE multipostion digital vortexer is a multipurpose shaker dedicated to a variety of aggressive shaking and mixing applications.
  • Compact design with easy-access control panel
  • Safe for cold room or incubator use
  • Five-year warranty
Three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform features spring-loaded retaining bars that adjust to fit vessels of different shapes and sizes.
The microplate platform isMais informação do produto