Print…
Cleanroom wipes, Anticon Guardrail® G2
Fornecedor: CONTEC
G2HW0101
CONTG2HW0101EA 689 EUR
CONTG2HW0101
Cleanroom wipes, Anticon Guardrail® G2
Toalhetes Toalhetes para salas limpas Seco
100% continuous filament polyester wipe with sealed border and Particle Attraction Technology (PAT).
  • Sealed border edge allows wipe to have low fibers and particles
  • Highly absorbent and abrasion resistant
  • Particle Attraction Technology (PAT) attracts 35 times more particles than other wipes
  • Excellent solvent compatibility with isopropyl alcohol, acetone, and other solvents
  • Ultra low levels of ions and extractables makes it ideal for critical cleaning tasks
Using Contec’s patented Particle Attraction Technology (PAT) system, the wipe attracts andMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Peso base 138,8 g/m²
Absorção - extrínseca 360,9 ml/m²
Absorção - intrínseca 2,60 ml/g
Absorção - índice 1 s
Extraíveis - usando água desionizada 0,005 g/m²
Extraíveis - usando álcool isopropílico 0,012 g/m²
Fibras (>100 µm) 0,692×10³/m²
Particle generation (>=0,5 µm) 18,5 × 10⁶/m²
Iões - cloreto 0,063 ppm
Iões - sódio 0,048 ppm