Print…

Anti-IgG2a Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Fornecedor: Columbia Biosciences
D5-112-2A
COBSD5-112-2AEA 385 EUR
COBSD5-112-2A
Anti-IgG2a Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Anticorpos
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR