Print…

S-Isopropylisothiouronium bromide

Fornecedor: Cayman Chemical

81290-100 81290-50 81290-500
CAYM81290-100EA 113 EUR
CAYM81290-100 CAYM81290-50 CAYM81290-500
S-Isopropylisothiouronium bromide
S-Isopropylisothiouronium bromide
Fórmula: C₄H₁₁BrN₂S
Número CAS: 57200-31-4

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR