Print…

Corticosterone ELISA Kits

Fornecedor: Cayman Chemical

501320-480 501320-96
CAYM501320-480EA 1870 EUR
CAYM501320-480 CAYM501320-96
Corticosterone ELISA Kits
Ensaios ELISAs

A competitive assay for the quantification of corticosterone in serum and rodent feces.


  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure corticosterone levels down to 30 pg/ml
  • Incubation: Overnight | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
  • Mid-point: 50% B/B0: 130 – 180 pg/ml


Corticosterone is a glucocorticoid produced by the adrenal cortex in response to ACTH (adrenocorticotropic hormone), and is the precursor to aldosterone. The production of glucorcorticoids is increased by stress; therefore, corticosterone can be used as a biomarker of stress. Plasma corticosterone levels have a circadian variation, and may be important in the regulation of the sleep-wake cycle. Cayman’s Corticosterone ELISA Kit is a competitive assay that can be used for quantification of corticosterone in serum and rodent feces.


Fornecimento: Kit contains: Antiserum, tracer, standard, buffers, reagents and plate.

Encomendar

ESPECIFICAÇÕES

Environmentally Preferable
Primary antibody reactivity
Tipo de ensaio Competitive
Hospedeiro
Sensibilidade 80% B/B0: 30 pg/ml
Faixa de detecção 8,2 - 5000 pg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR