Print…

Anti-S1P4 Rabbit Polyclonal Antibody

Fornecedor: Cayman Chemical
13489-1
CAYM13489-1EA 807 EUR
CAYM13489-1
Anti-S1P4 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: S1P4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR