Print…

Anti-CREB Rabbit Polyclonal Antibody

Fornecedor: Cayman Chemical
10009181-1
CAYM10009181-1EA 897 EUR
CAYM10009181-1
Anti-CREB Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR