Print…

Anti-PTEN Rabbit Polyclonal Antibody

Fornecedor: Cayman Chemical
10005059-500
CAYM10005059-500EA 582 EUR
CAYM10005059-500
Anti-PTEN Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Chimpanzee, Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR