Print…

Anti-Vtg Rabbit Polyclonal Antibody [clone: GA-306]

Fornecedor: Cayman Chemical
10004442-100 10004442-500
CAYM10004442-100EA 690 EUR
CAYM10004442-100 CAYM10004442-500
Anti-Vtg Rabbit Polyclonal Antibody [clone: GA-306]
Anticorpos
Type: Primary
Antigen: Vtg
Clonality: Polyclonal
Clone: GA-306
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: ballan Wrasse, Cod, Rainbow Trout, Sheepshead Minnow, three-spined stickleback, Tilapia, Turbot
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR