Measuring scoops
Supplier: BURKLE
9614-0005 9614-0050 9614-0010 9614-0500 9614-0025 9614-0100 9614-0250 9614-1000
BURK9614-0005EA 0.7 EUR
BURK9614-0005 BURK9614-0050 BURK9614-0010 BURK9614-0500 BURK9614-0025 BURK9614-0100 BURK9614-0250 BURK9614-1000
Measuring scoops
Scoops
Universal graduated PP shovels for filling and measuring of powders and granules. For exact metering, the graduated shovel is to be overfilled and the excess at the upper edge of the shovel is to be wiped off.
  • High chemical resistance
  • Calibrated volume when full to the brim
  • Suitable for food contact