Print…
Burette stopcocks, straight
Fornecedor: Brand
81937 81939
BRND81937EA 64.7 EUR
BRND81937 BRND81939
Burette stopcocks, straight
Torneiras
Borosilicate glass 5,4, amber.
  • Interchangeable key
  • With glass or PTFE key
  • Key with retention device
Stopcock housing with burette tip and key.