Print…
Label Printer, QWERTY EU, M610
Fornecedor: Brady
317797
BRDY317797EA 937 EUR
BRDY317797
Label Printer, QWERTY EU, M610
Impressoras Label Printers
Get the durability your work demands.
  • Design and print labels using a keypad or desktop software
  • Work longer with an enhanced Gen3 Li-ION battery – print 4500 labels on a full charge
  • Easily import and export label files, lists and graphics to the printer through USB 2.0 port (Type A and B) or USB flash drive
  • Print labels at 50,80 mm per second print speed
  • Get added durability with a printer that is resistant to military-grade shocks (MIL-STD-810G) and 1.80 m drops
  • Operate your printer in −18...+46 °C temperatures
Do more with enhanced capabilitiesMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Label Continuous and die-cut
Módulo Stand-alone and peripheral
Impressora Monocolour
Print method Thermal transfer
Print resolution 300 dpi
Print speed 750 (recommended usage per day)