Print…
Polyethylene Tags for M710, M610 and M611, Bulk
Fornecedor: Brady
312126
BRDY312126EA 326 EUR
BRDY312126
Polyethylene Tags for M710, M610 and M611, Bulk
Rótulos
Multipurpose tags can be used for a variety of applications including identification of multi-conductor cables, inventory, equipment, lockout, safety warning repair and work-in-progress. Extremely tear-resistant and superior cold-weather tag.
  • Cross-laminated polyethylene provides extreme tear resistance and excellent cold weather performance
  • Matte finish
Applications include data and telecommunications labelling, wire and cable labelling, inventory and inspection labelling, lockout tagout, and safety identification.
Embalagem: 1000 tags per box.