Print…
Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
Fornecedor: Brady
307652 307658 307653 307659
BRDY307652EA 7120 EUR
BRDY307652 116-0938 BRDY307653 BRDY307659
Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
Impressoras Label Printers
The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing on lots of different materials within the aerospace, defence and mass transit, electronics, automotive, logistics, electrical, voice-datacom and LAB markets.
  • Prints labels and then applies them to wires or cables
  • Two step process
  • Laboratory labelling application
  • Lithium-manganese battery
  • Colour: Monocolour
It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more. This printer comes withMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Ligações Ethernet, I/O connection, bluetooth (optional), WiFi (optional), serial, USB
Ecrã Colour touchscreen
Power supply AC adaptor
Print method Transfert thermique
Software Brady Workstation, Windows based driver for 3rd party software use
Supported OS Windows® 7, Windows® 8, Windows® 10, Windows® Server 2012, Windows® Server 2012 R2, Windows® Server 2016
Gama de temperaturas 5...40 °C