Print…
FreezerBondz™ Ultra-Thin Cryogenic Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Fornecedor: Brady
170778 170779 170899 170894 170780 M5-120-492 M5-117-492
BRDY170778EA 136 EUR
BRDY170778 BRDY170779 BRDY170899 BRDY170894 BRDY170780 BRDYM5-120-492 BRDYM5-117-492
FreezerBondz™ Ultra-Thin Cryogenic Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Rótulos
Label curved frozen lab surfaces including tubes, vials, and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady handheld label printer.
  • For use on smooth, curved surfaces
  • Withstands chemicals, abrasions, and extreme temperatures
  • Made with a permanent acrylic adhesive
Specialised labels are ideal for laboratory use on curved surfaces including test tubes, vials, and straw pipettes. Durable labels withstand chemicals, abrasions, and extreme temperatures, including storage in liquid nitrogen. TheMais informação do produto