Print…
Cable ties, stainless steel
Fornecedor: Brady
134057
BRDY134057EA 58.5 EUR
BRDY134057
Cable ties, stainless steel
Abraçadeiras
Brady B-7316 stainless steel cable ties are made from 316 grade stainless steel and are designed to withstand harsh environments, and offer excellent outdoor durability.
  • Thickness of 0,5 mm