Print…
Disposable safety scalpels, Aesculap®
Fornecedor: B. Braun
BA810SU BA811SU BA812SU BA815SU BA815-CSU BA818SU BA819SU BA820SU BA821SU BA822SU BA823SU BA824SU BA825SU BA836SU
BRAUBA810SUEA 27.1 EUR
BRAUBA810SU BRAUBA811SU BRAUBA812SU BRAUBA815SU BRAUBA815-CSU BRAUBA818SU BRAUBA819SU BRAUBA820SU BRAUBA821SU BRAUBA822SU BRAUBA823SU BRAUBA824SU BRAUBA825SU BRAUBA836SU
Disposable safety scalpels, Aesculap®
Bisturis Bisturis
Carbon steel blade with plastic handle and blade protector.
After use the blade can be securely locked. This minimises the risk of cutting incidents and prevents the scalpel from being used again.
Certificações: TRBA 250 certified