Print…
Three-fold multi-tap bank, Discofix® C
Fornecedor: B. Braun
16615C
BRAU16615CEA 1840 EUR
BRAU16615C
Three-fold multi-tap bank, Discofix® C
Infusões
Drug-Resistant Multi-Tap Systems are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, immunosuppressants and lipids.
  • Leaks caused by stress cracks, contamination and air embolism are prevented
  • Easy access through the rotary adapter for injections and aspirations
  • Control of the 45° position by 'clicking'