Print…
Stopcocks for desiccators
Fornecedor: Brand
82415
BRAA82415EA 61.7 EUR
BRAA82415
Stopcocks for desiccators
Torneiras
Stopcocks made of borosilicate glass 3.3 are suitable for Novus desiccators with lid socket.
  • Lateral venting helps to reduce turbulence
  • Nozzle with outer Ø 10 mm