Print…
Frascos de laboratório, gargalo estreito, com rosca
Fornecedor: Brand
704008 704010 704004 704006 704002 704020 704018 704016 704014 704012
BRAA704008EA 20.5 EUR
BRAA704008 BRAA704010 BRAA704004 BRAA704006 BRAA704002 BRAA704020 BRAA704018 BRAA704016 BRAA704014 BRAA704012
Frascos de laboratório, gargalo estreito, com rosca
Frascos Frascos de Vidro
Soda-lime glass, amber, coated or uncoated.
  • Good chemical and physical properties
  • Maximum operating temperature for coated bottles is 80 °C
  • To preserve the coating, do not clean at temperatures exceeding 60 °C
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber to protect light-sensitive materials. The coating envelops the glass bottle like a protection skin. Should the bottle break, the splintering hazards are reduced considerably.