Print…
Anti-BIN2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fornecedor: Bioss
BS-9727R-A350
BOSSBS-9727R-A350EA 893 EUR
BOSSBS-9727R-A350
Anti-BIN2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorpos
BAR proteins are characterized by a common N-terminal BAR (bin, amphiphysin and Rvs161/167) domain and are recognized as adaptor proteins that are involved in many cellular processes. BIN1 and BIN2 are BAR proteins that share 61% sequence similarity. BIN1 (Bridging integrator 1) is a ubiquitously expressed regulatory protein for synaptic vesicle endocytosis. BIN1 also interacts with the transcription factors c-Myc and MyoD, potentially functioning as a tumor suppressor. BIN2, also known as Breast cancer-associated protein 1, is a 565 amino acid protein that interacts with BIN1. In contrastMais informação do produto