Print…

Anti-MIEN1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornecedor: Bioss
BS-9636R-FITC
BOSSBS-9636R-FITCEA 900 EUR
BOSSBS-9636R-FITC
Anti-MIEN1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
Increases cell migration by inducing filopodia formation at the leading edge of migrating cells. Plays a role in regulation of apoptosis, possibly through control of CASP3. May be involved in a redox-related process (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MIEN1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR