Print…
Anti-ASB7 Rabbit Polyclonal Antibody
Fornecedor: Bioss
BS-9134R
BOSSBS-9134REA 725 EUR
BOSSBS-9134R
Anti-ASB7 Rabbit Polyclonal Antibody
Anticorpos
Members of the suppressor of cytokine signaling (SOCS) family of proteins contain C-terminal regions of homology called the SOCS box, which serves to couple SOCS proteins and their binding partners with the Elongin B/C complex. Several other families of proteins also contain SOCS boxes, but differ from the SOCS proteins in the type of domain they contain upstream of the SOCS box. The largest family of SOCS box-containing proteins is the ankyrin repeat and SOCS box-containing (ASB) protein family. ASB-7 (ankyrin repeat and SOCS box-containing 7) is a 318 amino acid protein that contains oneMais informação do produto