Print…

Anti-BARX1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Fornecedor: Bioss
BS-8521R-A647
BOSSBS-8521R-A647EA 893 EUR
BOSSBS-8521R-A647
Anti-BARX1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorpos
BarX1 belongs to the Bar subclass of the homeobox gene family. The function of this gene has not yet been determined; however, studies in the mouse and chick homolog suggest a role in developing teeth and craniofacial mesenchyme of neural crest origin. The role of these homologs implicates the human gene as a candidate for unmapped disorders involving tooth and jaw development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BARX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR