Print…
Anti-Azurocidin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fornecedor: Bioss
BS-7564R-A680
BOSSBS-7564R-A680EA 893 EUR
BOSSBS-7564R-A680
Anti-Azurocidin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorpos
This is a neutrophil granule-derived antibacterial and monocyte- and fibroblast-specific chemotactic glycoprotein. Binds heparin. The cytotoxic action is limited to many species of Gram-negative bacteria; this specificity may be explained by a strong affinity of the very basic N-terminal half for the negatively charged lipopolysaccharides that are unique to the Gram-negative bacterial outer envelope. It may play a role in mediating recruitment of monocytes in the second wave of inflammation. Has antibacterial activity against the Gram-nagative bacterium P.aeruginosa, this activity isMais informação do produto