Print…
Anti-RPL15 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fornecedor: Bioss
BS-6571R-A488
BOSSBS-6571R-A488EA 890 EUR
BOSSBS-6571R-A488
Anti-RPL15 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpos
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L15E family of ribosomal proteins. It is located in the cytoplasm. This gene shares sequence similarity with the yeast ribosomal protein YL10 gene. Although this gene has been referred to as RPL10, its official symbol is RPL15. This gene has been shown to beMais informação do produto