Print…

Anti-MPO Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Fornecedor: Bioss
BS-4943R-A750
BOSSBS-4943R-A750EA 899 EUR
BOSSBS-4943R-A750
Anti-MPO Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorpos
Part of the host defence system of polymorphonuclear leukocytes. It is responsible for microbicidal activity against a wide range of organisms. In the stimulated PMN, MPO catalyses the production of hypohalous acids, primarily hypochlorous acid in physiologic situations, and other toxic intermediates that greatly enhance PMN microbicidal activity.

Type: Primary
Antigen: MPO
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 678-745/745
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR