Print…
Anti-IRGM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornecedor: Bioss
BS-4015R-HRP
BOSSBS-4015R-HRPEA 900 EUR
BOSSBS-4015R-HRP
Anti-IRGM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
IRGM belongs to a family of interferon-gamma inducible GTPases, belonging to the GTPase superfamily, and are selectively induced by IFN-gamma or bacterial lipopolysaccharide (LPS) stimulation. IRGM is primarily expressed in all cells derived from B-cell lineages, and is highly expressed in macrophages following IFN-gamma stimulation. IRGM has been shown to be required for host defences against a broad range of intracellular pathogens. Specifically, IRGM deficient mice show defects with intracellular microbial killing, phagosome maturation and autophagy. More recently IRGM has been shown toMais informação do produto