Print…

Anti-MDH1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornecedor: Bioss
BS-3996R-A555
BOSSBS-3996R-A555EA 876 EUR
BOSSBS-3996R-A555
Anti-MDH1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpos
Malate dehydrogenase catalyzes the reversible oxidation of malate to oxaloacetate, utilizing the NAD/NADH cofactor system in the citric acid cycle. Malate dehydrogenase 1 (MDH1) is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR