Print…

Anti-SPINK1 Rabbit Polyclonal Antibody (Cy7®)

Fornecedor: Bioss
BS-2917R-CY7
BOSSBS-2917R-CY7EA 900 EUR
BOSSBS-2917R-CY7
Anti-SPINK1 Rabbit Polyclonal Antibody (Cy7®)
Anticorpos
This is a trypsin inhibitor, its physiological function is to prevent the trypsin-catalyzed premature activation of zymogens within the pancreas.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPINK1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR