Print…

Anti-CISD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Fornecedor: Bioss
BS-13959R-A750
BOSSBS-13959R-A750EA 890 EUR
BOSSBS-13959R-A750
Anti-CISD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorpos
Plays a key role in regulating maximal capacity for electron transport and oxidative phosphorylation (By similarity). May be involved in Fe-S cluster shuttling and/or in redox reactions.

Type: Primary
Antigen: CISD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-108/108
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR