Print…
Anti-LIMS2 Rabbit Polyclonal Antibody (Cy5®)
Fornecedor: Bioss
BS-13650R-CY5
BOSSBS-13650R-CY5EA 900 EUR
BOSSBS-13650R-CY5
Anti-LIMS2 Rabbit Polyclonal Antibody (Cy5®)
Anticorpos
Pinch is a focal adhesion protein that is a component of the ILK-PINCH complex. This complex is a major part of the growth factor and integrin signaling pathway. Pinch is involved in cell differentiation, proliferation and survival by acting as an effector of integrin and growth factor signaling. It is a cytoplasmic protein expressed in most tissues and consists of five LIM domains, a nuclear localization signal and a nuclear export signal. The PINCH-1/ILK complex is regulated by another member of the Pinch family, PINCH-2, which also forms a complex with ILK.
Recommended Dilutions: IF(Mais informação do produto