Print…
Anti-MBOAT4 Rabbit Polyclonal Antibody (Cy3®)
Fornecedor: Bioss
BS-13355R-CY3
BOSSBS-13355R-CY3EA 909 EUR
BOSSBS-13355R-CY3
Anti-MBOAT4 Rabbit Polyclonal Antibody (Cy3®)
Anticorpos
MBOAT4 (membrane-bound O-acyltransferase domain-containing protein 4, Ghrelin O-acyltransferase) is a 435 amino acid, multi-pass membrane protein that belongs to the membrane-bound acyltransferase family. MBOAT4 functions as an enzyme that attaches an octanoate fatty acid to serine-3 of ghrelin. Ghrelin is a very small, appetite-stimulating hormone secreted by the food-deprived stomach. MBOAT4 can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid. MBOAT4 expression, consistent with its function, is mainly in the stomach and intestines.Mais informação do produto