Print…

Anti-SHISA9 Rabbit Polyclonal Antibody

Fornecedor: Bioss
BS-11950R
BOSSBS-11950REA 725 EUR
BOSSBS-11950R
Anti-SHISA9 Rabbit Polyclonal Antibody
Anticorpos
Regulator of short-term neuronal synaptic plasticity in the dentate gyrus. Associates with AMPA receptors (ionotropic glutamate receptors) in synaptic spines and promotes AMPA receptor desensitization at excitatory synapses.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SHISA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

ESPECIFICAÇÕES

Símbolo do antígeno SHISA9
Nome do antigénio Shisa family member 9
Environmentally Preferable
Sinônimos de antígeno SHISA9|Protein shisa-9|CKAMP44|Cytosine-knot AMPA Receptor modulating Protein 44a
Tipo de anticorpo Primário
Clonalidade Policlonal
Conjugado Não conjugado
Reatividade Human,Rat,Mouse
Hospedeiro Rabbit
Isótipo IgG
Western blot Yes
ImunoQuímica Yes
Forma liquid
Purification Purified by Protein A.
Concentração 1 ug/ul
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR