Print…

Anti-S1PR5 Rabbit Polyclonal Antibody (Cy5®)

Fornecedor: Bioss
BS-11457R-CY5
BOSSBS-11457R-CY5EA 900 EUR
BOSSBS-11457R-CY5
Anti-S1PR5 Rabbit Polyclonal Antibody (Cy5®)
Anticorpos
Receptor for the lysosphingolipid sphingosine 1-phosphate (S1P). S1P is a bioactive lysophospholipid that elicits diverse physiological effect on most types of cells and tissues. Is coupled to both the G(i/0)alpha and G(12) subclass of heteromeric G-proteins (By similarity). May play a regulatory role in the transformation of radial glial cells into astrocytes and may affect proliferative activity of these cells.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: S1PR5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Mais informação do produto
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR