Print…
Anti-Gamma-taxilin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornecedor: Bioss
BS-0631R-FITC
BOSSBS-0631R-FITCEA 900 EUR
BOSSBS-0631R-FITC
Anti-Gamma-taxilin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
This gene encodes a member of the taxilin family. The encoded protein binds to the C-terminal coiled-coil region of syntaxin family members 1A, 3A and 4A, and may play a role in intracellular vesicle trafficking. This gene is up-regulated by lipopolysaccharide and the gene product may be involved in cell cycle regulation. The related mouse protein was also shown to inhibit activating transcription factor 4-mediated transcription and thus regulate bone mass accrual. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Dec 200Mais informação do produto