Print…
Cover spectacles, Tryon OTG
Fornecedor: Bollé Safety
TRYOTGPSI TRYOTGPSF
BOLETRYOTGPSIEA 19.1 EUR
BOLETRYOTGPSI BOLETRYOTGPSF
Cover spectacles, Tryon OTG
Óculos
These Premium OTG features provide maximum coverage and extendable temples for a precise fit. The Tryon OTG can be worn with most prescription spectacles.
  • Flexible upper forehead protection (TPR)
  • PC/TPR adjustable and stretchable co-injected temples
  • Temples FLEX 160°
  • Wash resistant (water or soap)
  • High speed particles at extreme temperatures
  • Low energy impact
  • 99,9% UVA/UVB protection
The permanent PLATINUM® coating applied on both sides of the lens makes it highly scratch-resistant and durably delays the appearance of fogging which guarantees evenMais informação do produto