Print…
Mini-display splitter, b.safe
Fornecedor: Bohlender
R658-04 R659-04
BOHLR658-04EA 301 EUR
BOHLR658-04 BOHLR659-04
Mini-display splitter, b.safe
Sensores de nível
Display unit to control the b.safe Splitter, which is available as an option in combination with connected fill-level sensors in b.safe Waste Caps and b.safe System Waste Caps.
  • With plugs for EU, Australia, USA, Japan, and UK
  • Compatible with any version of b.safe Waste Caps
  • Additional comfort provides the possibility to snooze the alarm tone during the canister exchange
Alarm box with touch buttons and LED indicators to monitor the operation and the fill level of the connected sensors. Programmed only to control the b.safe Splitter for monitoring 2Mais informação do produto