Print…
b.safe Air Valve PFAS Analytic
Fornecedor: Bohlender
M405-01
BOHLM405-01EA 50.8 EUR
BOHLM405-01
b.safe Air Valve PFAS Analytic
Válvulas Válvulas de regulação de ar
b.safe Air Valve PFAS Analytic, free from polyhaloolefins, suitable for PFAS analysis without restrictions.
  • Valve housing made of PP with connection (thread UNF ¹/₄" 28G) suitable for b.safe caps
  • Cover with ventilation opening and luerlock connection
  • Integrated check valve
  • Upstream diaphragm made of PP (1 μm)
  • Including attachable change indicator
For extraction of eluents in context of the PFAS analytic. The b.safe Air Valve is especially adapted to the flow rate in HPLC. The integrated non-return valve avoids evaporation of solvents and lets flowMais informação do produto