Print…
Stoppers, PTFE
Fornecedor: Bohlender
H936-03 H937-04 H937-05 H936-04 H936-07 H937-07 H936-06 H936-05 H936-10 H936-02 H937-10 H936-08
BOHLH936-03EA 11.5 EUR
BOHLH936-03 BOHLH937-04 BOHLH937-05 BOHLH936-04 BOHLH936-07 BOHLH937-07 BOHLH936-06 BOHLH936-05 BOHLH936-10 BOHLH936-02 BOHLH937-10 BOHLH936-08
Stoppers, PTFE
Rolhas Rolhas de PE/LDPE/HDPE
For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are easily removable and can be used without grease.
  • PTFE
  • Temperature resistance from −200 to + 250 °C
  • Chemical resistance
  • FDA conform
The stoppers can expand under heat which might lead to a breaking of the ground joint sockets.