Print…
Stopcocks, miniature
Fornecedor: Bohlender
F731-02 F731-04 F731-06 F731-08 F730-04 F730-06 F731-16 F731-14 F731-12 F731-10 F730-08 F730-02
BOHLF731-02EA 111 EUR
BOHLF731-02 BOHLF731-04 BOHLF731-06 BOHLF731-08 BOHLF730-04 BOHLF730-06 BOHLF731-16 BOHLF731-14 BOHLF731-12 BOHLF731-10 BOHLF730-08 BOHLF730-02
Stopcocks, miniature
Torneiras
Miniature stopcocks of PTFE with male or female connection threads and stopcock plug of FEP.
  • Temperature resistance from -200 to 205°C
  • Universally chemical resistant
  • Pressure upto 8 bar