Print…
Conchas de farmácia
Fornecedor: BOCHEM
8810 8811 8812 8813 8809
BOCH8810EA 168 EUR
BOCH8810 231-0205 BOCH8812 BOCH8813 BOCH8809
Conchas de farmácia
Conchas
These chemical scoops made from stainless steel 18/10 are ideal for use in pharmaceutical and food industries.
  • With welded handle
  • Food contact approved